Gene Transcripts
- Gene Symbol
- C15orf41
- Entrez Gene ID
- 84529
- Gene Name
- chromosome 15 open reading frame 41
- Gene Aliases
- HH114
- Chromosome Location
- Chr. 15: 36579603 - 36810260 on Build GRCh38
- UniGene ID
- Hs.48348
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130010.2 |
NP_001123482.1
|
6, 7 |
683 |
|
NM_001290232.1 |
NP_001277161.1
|
6, 7 |
374 |
|
NM_001290233.1 |
NP_001277162.1
|
6, 7 |
683 |
|
NM_001321756.1 |
NP_001308685.1
|
5, 6 |
637 |
|
NM_001321757.1 |
NP_001308686.1
|
7, 8 |
717 |
|
NM_001321758.1 |
NP_001308687.1
|
4, 5 |
572 |
|
NM_001321759.1 |
NP_001308688.1
|
6, 7 |
683 |
|
NM_001321760.1 |
NP_001308689.1
|
6, 7 |
683 |
|
NM_001321761.1 |
NP_001308690.1
|
6, 7 |
683 |
|
NM_032499.5 |
NP_115888.1
|
7, 8 |
429 |
|
XM_011522111.1 |
XP_011520413.1
|
6, 7 |
675 |
|
GenBank mRNA |
AF114263.1 |
- |
7, 8 |
404 |
|
AK026504.1 |
- |
6, 7 |
652 |
|
AK315446.1 |
- |
6, 7 |
343 |
|
BC006254.2 |
- |
6, 7 |
442 |
3952242 |
View Full Product Details