Gene Transcripts

Gene Symbol
LTV1
Entrez Gene ID
84946
Gene Name
LTV1 ribosome biogenesis factor
Gene Aliases
C6orf93, dJ468K18.4
Chromosome Location
Chr. 6: 143843371 - 143863806 on Build GRCh38
UniGene ID
Hs.185675
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032860.3 NP_116249.2 2 228
GenBank mRNA AK095269.1 - 2 259
AK300360.1 - 2 214
BC009855.2 - 2 228 3926227

View Full Product Details