Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033100.3 | NP_149091.1 | 17 | 4765 | |
XM_011540337.1 | XP_011538639.1 | 16 | 5256 | ||
XM_011540339.1 | XP_011538641.1 | 17 | 4571 | ||
GenBank mRNA | AB593155.1 | - | 17 | 4751 | |
AK124752.1 | - | 10 | 3854 | ||
AL080188.1 | - | 2 | 3071 |