Gene Transcripts
- Gene Symbol
- MYB
- Entrez Gene ID
- 4602
- Gene Name
- MYB proto-oncogene, transcription factor
- Gene Aliases
- Cmyb, c-myb, c-myb_CDS, efg
- Chromosome Location
- Chr. 6: 135180981 - 135219173 on Build GRCh38
- UniGene ID
- Hs.606320
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130172.1 |
NP_001123644.1
|
2, 3 |
346 |
|
NM_001130173.1 |
NP_001123645.1
|
2, 3 |
346 |
|
NM_001161656.1 |
NP_001155128.1
|
2, 3 |
346 |
|
NM_001161657.1 |
NP_001155129.1
|
2, 3 |
346 |
|
NM_001161658.1 |
NP_001155130.1
|
2, 3 |
346 |
|
NM_001161659.1 |
NP_001155131.1
|
2, 3 |
346 |
|
NM_001161660.1 |
NP_001155132.1
|
2, 3 |
346 |
|
NM_005375.3 |
NP_005366.2
|
2, 3 |
346 |
|
NR_134958.1 |
|
2, 3 |
346 |
|
NR_134959.1 |
|
2, 3 |
346 |
|
NR_134960.1 |
|
2, 3 |
346 |
|
NR_134961.1 |
|
2, 3 |
346 |
|
NR_134962.1 |
|
2, 3 |
346 |
|
NR_134963.1 |
|
2, 3 |
346 |
|
NR_134964.1 |
|
2, 3 |
346 |
|
NR_134965.1 |
|
2, 3 |
346 |
|
GenBank mRNA |
AF104863.1 |
- |
2, 3 |
224 |
|
AJ606317.1 |
- |
2, 3 |
346 |
|
AJ606318.1 |
- |
2, 3 |
346 |
|
AJ606319.1 |
- |
2, 3 |
346 |
|
AJ606320.1 |
- |
2, 3 |
346 |
|
AJ606321.1 |
- |
2, 3 |
346 |
|
AJ606322.1 |
- |
2, 3 |
346 |
|
AJ606323.1 |
- |
2, 3 |
346 |
|
AJ606324.1 |
- |
2, 3 |
346 |
|
AJ616235.1 |
- |
2, 3 |
261 |
|
AY787443.1 |
- |
2, 3 |
147 |
|
AY787446.1 |
- |
2, 3 |
147 |
|
AY787447.1 |
- |
2, 3 |
147 |
|
AY787448.1 |
- |
2, 3 |
147 |
|
AY787449.1 |
- |
2, 3 |
147 |
|
AY787450.1 |
- |
2, 3 |
147 |
|
AY787451.1 |
- |
2, 3 |
147 |
|
AY787452.1 |
- |
2, 3 |
147 |
|
AY787453.1 |
- |
2, 3 |
147 |
|
AY787454.1 |
- |
2, 3 |
147 |
|
AY787455.1 |
- |
2, 3 |
147 |
|
AY787456.1 |
- |
2, 3 |
147 |
|
AY787457.1 |
- |
2, 3 |
147 |
|
AY787458.1 |
- |
2, 3 |
147 |
|
AY787459.1 |
- |
2, 3 |
147 |
|
AY787460.1 |
- |
2, 3 |
147 |
|
AY787461.1 |
- |
2, 3 |
147 |
|
AY787462.1 |
- |
2, 3 |
147 |
|
AY787463.1 |
- |
2, 3 |
147 |
|
AY787464.1 |
- |
2, 3 |
147 |
|
AY787465.1 |
- |
2, 3 |
147 |
|
AY787466.1 |
- |
2, 3 |
147 |
|
AY787467.1 |
- |
2, 3 |
147 |
|
AY787468.1 |
- |
2, 3 |
147 |
|
AY787469.1 |
- |
2, 3 |
147 |
|
AY787470.1 |
- |
2, 3 |
147 |
|
AY787471.1 |
- |
2, 3 |
147 |
|
AY787472.1 |
- |
2, 3 |
147 |
|
AY787473.1 |
- |
2, 3 |
147 |
|
AY787474.1 |
- |
2, 3 |
147 |
|
AY787475.1 |
- |
2, 3 |
147 |
|
BC064955.1 |
- |
2, 3 |
298 |
6069320 |
M15024.1 |
- |
2, 3 |
260 |
|
X52125.1 |
- |
2, 3 |
344 |
|
X52126.1 |
- |
1, 2 |
150 |
|
View Full Product Details