Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005583.4 | NP_005574.2 | 3 | 1014 | |
XM_005259912.3 | XP_005259969.1 | 3 | 852 | ||
XM_017026816.1 | XP_016882305.1 | 4 | 1714 | ||
XM_017026817.1 | XP_016882306.1 | 4 | 1567 | ||
GenBank mRNA | BC002796.2 | - | 3 | 721 | |
M22637.1 | - | 3 | 579 |