Gene Transcripts

Gene Symbol
Atpif1
Entrez Gene ID
25392
Gene Name
ATPase inhibitory factor 1
Gene Aliases
Atpi, IF1PA
Chromosome Location
Chr. 5: 150719570 - 150723287 on Build Rnor_6.0
UniGene ID
Rn.1658
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012915.2 NP_037047.2 2 198
GenBank mRNA BC126065.1 - 2 163
D13122.1 - 2 198
FQ221416.1 - 2 183
FQ229412.1 - 2 160

View Full Product Details