Gene Transcripts

Gene Symbol
Htr6
Entrez Gene ID
64354
Gene Name
5-hydroxytryptamine (serotonin) receptor 6, G protein-coupled
Gene Aliases
Chromosome Location
Chr. 5: 157503040 - 157518511 on Build Rnor_6.0
UniGene ID
Rn.10552
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024365.1 NP_077341.1 3 1460
GenBank mRNA L03202.1 - 3 1577
L19656.1 - 3 1460
L41146.1 - 3 1143
S62043.1 - 3 1460

View Full Product Details