Gene Transcripts

Gene Symbol
Prss3
Entrez Gene ID
286911
Gene Name
protease, serine, 3
Gene Aliases
Try3
Chromosome Location
Chr. 4: 69264628 - 69268336 on Build Rnor_6.0
UniGene ID
Rn.9817
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173127.1 NP_775150.1 2 192
GenBank mRNA FQ226997.1 - 2 223
FQ230379.1 - 2 223
FQ235277.1 - 2 223
M16624.1 - 2 192

View Full Product Details