Gene Transcripts

Gene Symbol
Atp1a3
Entrez Gene ID
24213
Gene Name
ATPase, Na+/K+ transporting, alpha 3 polypeptide
Gene Aliases
Atpa1a3
Chromosome Location
Chr. 1: 81852425 - 81881549 on Build Rnor_6.0
UniGene ID
Rn.87329
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012506.1 NP_036638.1 6 644
XM_008758888.1 XP_008757110.1 6 558
GenBank mRNA FQ212331.1 - 5 481
M14513.1 - 6 644
X05883.1 - 6 641

View Full Product Details