Gene Transcripts

Gene Symbol
Naf1
Entrez Gene ID
234344
Gene Name
nuclear assembly factor 1 homolog (S. cerevisiae)
Gene Aliases
Gm174
Chromosome Location
Chr. 8: 66859957 - 66890564 on Build GRCm38
UniGene ID
Mm.334561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163564.1 NP_001157036.1 8 1629
GenBank mRNA AK144735.1 - 8 876
BC027148.1 - 4 412
BC053440.1 - 4 400

View Full Product Details