Gene Transcripts

Gene Symbol
Npat
Entrez Gene ID
244879
Gene Name
nuclear protein in the AT region
Gene Aliases
6820401K01, AI427561, BB112559
Chromosome Location
Chr. 9: 53537041 - 53575627 on Build GRCm38
UniGene ID
Mm.116713
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081152.1 NP_001074621.1 8 805
GenBank mRNA AK033012.1 - 8 785
AK080294.1 - 8 806

View Full Product Details