Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028440.1 | NP_082716.1 | 3 | 528 | |
GenBank mRNA | AK013984.1 | - | 3 | 490 | |
AK171138.1 | - | 3 | 431 | ||
BC039801.1 | - | 3 | 407 | ||
BC098509.1 | - | 3 | 470 | ||
DV053786.1 | - | 3 | 577 |