Gene Transcripts
- Gene Symbol
- Myo18b
- Entrez Gene ID
- 74376
- Gene Name
- myosin XVIIIb
- Gene Aliases
- 4932408L24Rik, 4933411E19Rik, Gm448
- Chromosome Location
- Chr. 5: 112688876 - 112896459 on Build GRCm38
- UniGene ID
- Mm.441903
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_028901.2 |
NP_083177.2
|
4 |
1215 |
|
View Full Product Details