Gene Transcripts
- Gene Symbol
- N5
- Entrez Gene ID
- 64825
- Gene Name
- DNA binding protein N5
- Gene Aliases
- Chromosome Location
- Chr. 4: 57588724 - 57590923 on Build Rnor_6.0
- UniGene ID
- Rn.202953
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022857.1 |
NP_074048.1
|
2 |
1287 |
|
GenBank mRNA |
L31882.1 |
- |
2 |
1287 |
|
View Full Product Details