Gene Transcripts
- Gene Symbol
- Abi1
- Entrez Gene ID
- 79249
- Gene Name
- abl-interactor 1
- Gene Aliases
- E3b1
- Chromosome Location
- Chr. 17: 89951664 - 90033334 on Build Rnor_6.0
- UniGene ID
- Rn.43675
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024397.2 |
NP_077373.2
|
1, 2 |
189 |
|
XM_006254353.2 |
XP_006254415.1
|
1, 2 |
260 |
|
XM_006254354.2 |
XP_006254416.1
|
1, 2 |
259 |
|
XM_006254355.2 |
XP_006254417.1
|
1, 2 |
259 |
|
XM_006254356.2 |
XP_006254418.1
|
1, 2 |
259 |
|
XM_006254357.2 |
XP_006254419.1
|
1, 2 |
259 |
|
XM_006254358.2 |
XP_006254420.1
|
1, 2 |
259 |
|
XM_006254359.2 |
XP_006254421.1
|
1, 2 |
259 |
|
XM_006254360.2 |
XP_006254422.1
|
1, 2 |
277 |
|
GenBank mRNA |
AF176784.1 |
- |
1, 2 |
126 |
|
BC129092.1 |
- |
1, 2 |
145 |
7192971 |
View Full Product Details