Gene Transcripts

Gene Symbol
Scrg1
Entrez Gene ID
64458
Gene Name
stimulator of chondrogenesis 1
Gene Aliases
Chromosome Location
Chr. 16: 36127536 - 36161041 on Build Rnor_6.0
UniGene ID
Rn.7724
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033499.1 NP_277034.1 2 248
GenBank mRNA AJ132434.1 - 2 248

View Full Product Details