Gene Transcripts

Gene Symbol
Pex11a
Entrez Gene ID
85249
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
Pmp26p
Chromosome Location
Chr. 1: 141474189 - 141481270 on Build Rnor_6.0
UniGene ID
Rn.14519
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053487.2 NP_445939.1 3 537
XM_008759560.1 XP_008757782.1 3 536
GenBank mRNA AJ224120.1 - 3 569

View Full Product Details