Gene Transcripts

Gene Symbol
Slc5a7
Entrez Gene ID
85426
Gene Name
solute carrier family 5 (sodium/choline cotransporter), member 7
Gene Aliases
Cht1
Chromosome Location
Chr. 9: 5293381 - 5330976 on Build Rnor_6.0
UniGene ID
Rn.44872
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053521.1 NP_445973.1 3 424
XM_006244251.2 XP_006244313.1 3 527
XM_006244253.2 XP_006244315.1 1 71
XM_008766774.1 XP_008764996.1 3 575
XM_008766775.1 XP_008764997.1 3 524
GenBank mRNA AB030947.1 - 3 424

View Full Product Details