Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144026.2 | NP_001137498.1 | 13 | 1772 | |
NM_001144028.2 | NP_001137500.1 | 13 | 1724 | ||
NM_014434.3 | NP_055249.1 | 13 | 1745 | ||
XM_011518545.1 | XP_011516847.1 | 12 | 1660 | ||
XM_011518547.1 | XP_011516849.1 | 13 | 1562 | ||
XM_011518548.2 | XP_011516850.1 | 12 | 2962 | ||
GenBank mRNA | AF199509.1 | - | 13 | 1681 | |
AK026089.1 | - | 13 | 1744 | ||
AK225995.1 | - | 13 | 1743 | ||
AK290026.1 | - | 13 | 1742 | ||
AY077845.1 | - | 13 | 1690 | ||
BC015735.1 | - | 13 | 1681 | 4869561 | |
BC078668.1 | - | 13 | 1681 | ||
BC093782.1 | - | 13 | 1662 | 7939627 | |
BC111943.1 | - | 13 | 1662 | 8327411 |