Gene Transcripts

Gene Symbol
Elovl5
Entrez Gene ID
171400
Gene Name
ELOVL fatty acid elongase 5
Gene Aliases
rELO1
Chromosome Location
Chr. 8: 85220947 - 85287449 on Build Rnor_6.0
UniGene ID
Rn.212229
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134382.1 NP_599209.1 2 195
XM_006243417.1 XP_006243479.1 3 360
GenBank mRNA AB071985.1 - 2 195

View Full Product Details