Gene Transcripts

Gene Symbol
Arfgef2
Entrez Gene ID
296380
Gene Name
ADP-ribosylation factor guanine nucleotide-exchange factor 2 (brefeldin A-inhibited)
Gene Aliases
Big2
Chromosome Location
Chr. 3: 163570437 - 163656612 on Build Rnor_6.0
UniGene ID
Rn.161866
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181083.2 NP_851597.2 10, 11 1454
XM_006235640.2 XP_006235702.1 10, 11 1636
GenBank mRNA AY255526.2 - 10, 11 1454

View Full Product Details