Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133286.1 | NP_579820.1 | 4 | 230 | |
XM_006231436.2 | XP_006231498.1 | 4 | 422 | ||
XM_006231437.2 | XP_006231499.1 | 5 | 350 | ||
XM_008760398.1 | XP_008758620.1 | 5 | 317 | ||
GenBank mRNA | AB079113.1 | - | 4 | 230 |