Gene Transcripts

Gene Symbol
Cdh6
Entrez Gene ID
12563
Gene Name
cadherin 6
Gene Aliases
K-cadherin, cad6, cadherin-6
Chromosome Location
Chr. 15: 13028699 - 13173689 on Build GRCm38
UniGene ID
Mm.57048
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007666.3 NP_031692.2 12 2423
GenBank mRNA BC107008.1 - 11 2164
BC107013.1 - 11 2164 40048308
D82029.1 - 11 2164

View Full Product Details