Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136059.2 | NP_001129531.1 | 2 | 799 | |
NM_009992.4 | NP_034122.1 | 2 | 878 | ||
GenBank mRNA | AK005000.1 | - | 2 | 879 | |
BC125440.1 | - | 2 | 798 | 40129955 | |
BC125444.1 | - | 2 | 798 | 40129959 | |
K02588.1 | - | 2 | 798 | ||
Y00071.1 | - | 2 | 799 |