Gene Transcripts

Gene Symbol
Sqstm1
Entrez Gene ID
18412
Gene Name
sequestosome 1
Gene Aliases
A170, OSF-6, Osi, STAP, STONE14, p62
Chromosome Location
Chr. 11: 50200152 - 50210820 on Build GRCm38
UniGene ID
Mm.40828
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290769.1 NP_001277698.1 3 553
NM_011018.3 NP_035148.1 3 553
GenBank mRNA BC006019.1 - 3 550
E08146.1 - 3 511
U40930.1 - 3 523
U57413.1 - 3 515

View Full Product Details