Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278734.1 | NP_001265663.1 | 8 | 2791 | |
NM_016324.3 | NP_057408.2 | 5 | 2475 | ||
NM_016325.3 | NP_057409.1 | 7 | 2680 | ||
NM_133502.2 | NP_598009.1 | 8 | 2776 | ||
XM_011526327.1 | XP_011524629.1 | 7 | 2214 | ||
XM_011526328.2 | XP_011524630.1 | 3 | 1895 | ||
XM_017026174.1 | XP_016881663.1 | 6 | 2481 | ||
XM_017026175.1 | XP_016881664.1 | 6 | 2636 | ||
GenBank mRNA | AB029149.1 | - | 5 | 2435 | |
AB029150.1 | - | 7 | 2582 | ||
AF275680.1 | - | 8 | 2455 | ||
AF275681.1 | - | 2 | 1889 | ||
AK095162.1 | - | 2 | 3120 | ||
AK222878.1 | - | 5 | 2403 | ||
BC009763.2 | - | 7 | 2303 | 2966764 | |
CA942346.1 | - | 1 | 410 |