Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011482.4 | NP_035612.2 | 3 | 442 | |
GenBank mRNA | AK004489.1 | - | 3 | 390 | |
AK134943.1 | - | 380 | |||
AK145391.1 | - | 3 | 444 | ||
AK145608.1 | - | 3 | 424 | ||
AK168808.1 | - | 3 | 412 | ||
AK168930.1 | - | 3 | 395 | ||
AK169103.1 | - | 3 | 426 | ||
BC026755.1 | - | 3 | 388 | 3154066 | |
BC054450.1 | - | 3 | 388 | 6491298 | |
BC083315.1 | - | 3 | 364 | 5714819 | |
U95114.1 | - | 3 | 415 |