Gene Transcripts
- Gene Symbol
- Rabep1
- Entrez Gene ID
- 54189
- Gene Name
- rabaptin, RAB GTPase binding effector protein 1
- Gene Aliases
- Rab5ep, rabaptin-5, rabaptin-5alpha
- Chromosome Location
- Chr. 11: 70844723 - 70943105 on Build GRCm38
- UniGene ID
- Mm.7087
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291142.1 |
NP_001278071.1
|
2, 3 |
384 |
|
NM_001291143.1 |
NP_001278072.1
|
3, 4 |
455 |
|
NM_019400.3 |
NP_062273.2
|
2, 3 |
384 |
|
GenBank mRNA |
AB001750.1 |
- |
3, 4 |
291 |
|
AF248489.1 |
- |
2, 3 |
170 |
|
AK019413.1 |
- |
2, 3 |
358 |
|
AK134587.1 |
- |
2, 3 |
383 |
|
AK137236.1 |
- |
2, 3 |
349 |
|
AK151901.1 |
- |
2, 3 |
353 |
|
BC009814.1 |
- |
2, 3 |
380 |
|
BC060166.1 |
- |
2, 3 |
363 |
|
BC080274.1 |
- |
2, 3 |
357 |
|
BC129853.1 |
- |
2, 3 |
203 |
40109688 |
BC129854.1 |
- |
2, 3 |
203 |
40109689 |
BY718394.1 |
- |
2, 3 |
383 |
|
D86066.1 |
- |
2, 3 |
220 |
|
View Full Product Details