Gene Transcripts
- Gene Symbol
- Tspan3
- Entrez Gene ID
- 56434
- Gene Name
- tetraspanin 3
- Gene Aliases
- 1700055K04Rik, TM4-A, Tm4sf8
- Chromosome Location
- Chr. 9: 56135884 - 56161070 on Build GRCm38
- UniGene ID
- Mm.28484
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019793.3 |
NP_062767.3
|
3, 4 |
546 |
|
GenBank mRNA |
AB009021.1 |
- |
3, 4 |
328 |
|
AB072450.1 |
- |
3, 4 |
356 |
|
AF133427.1 |
- |
3, 4 |
340 |
|
AF242591.1 |
- |
3, 4 |
539 |
|
AK006798.1 |
- |
3, 4 |
422 |
|
AK053325.1 |
- |
3, 4 |
397 |
|
AK089494.1 |
- |
|
378 |
|
AK141951.1 |
- |
3, 4 |
404 |
|
AK146130.1 |
- |
3, 4 |
534 |
|
AK152175.1 |
- |
3, 4 |
431 |
|
AK154919.1 |
- |
|
378 |
|
AK159324.1 |
- |
3, 4 |
517 |
|
AK159418.1 |
- |
3, 4 |
490 |
|
AK159621.1 |
- |
3, 4 |
501 |
|
AK159914.1 |
- |
3, 4 |
479 |
|
AK168231.1 |
- |
3, 4 |
534 |
|
AK169068.1 |
- |
3, 4 |
517 |
|
AK178223.1 |
- |
1, 2 |
25 |
|
AK180027.1 |
- |
1, 2 |
24 |
|
AK182665.1 |
- |
1, 2 |
24 |
|
AK187762.1 |
- |
|
11 |
|
AK207599.1 |
- |
|
24 |
|
AK220124.1 |
- |
1, 2 |
24 |
|
BC043072.1 |
- |
3, 4 |
524 |
6404165 |
View Full Product Details