Gene Transcripts

Gene Symbol
Habp4
Entrez Gene ID
56541
Gene Name
hyaluronic acid binding protein 4
Gene Aliases
4933413D03Rik, 4933428J01Rik
Chromosome Location
Chr. 13: 64161866 - 64186537 on Build GRCm38
UniGene ID
Mm.40989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019986.3 NP_064370.2 2 513
GenBank mRNA AF227684.1 - 2 456
AK016800.1 - 2 556
AK144206.1 - 2 497
BC082806.1 - 2 515

View Full Product Details