Gene Transcripts

Gene Symbol
Rpl35a
Entrez Gene ID
57808
Gene Name
ribosomal protein L35A
Gene Aliases
2810431L15Rik, Rpl35
Chromosome Location
Chr. 16: 33056453 - 33060189 on Build GRCm38
UniGene ID
Mm.19355
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130484.1 NP_001123956.1 4 399
NM_001130485.1 NP_001123957.1 4 383
NM_021338.3 NP_067313.2 4 429
GenBank mRNA AK003063.1 - 4 353
AK010692.1 - 4 350
AK028214.1 - 4 353
AK046386.1 - 4 355
AK160963.1 - 3 1491
BB611247.1 - 4 351
BC027223.1 - 4 398 4036717
BC086771.1 - 3 336
BC090255.1 - 3 336 2655001
BC100332.1 - 320 30846392
BC100599.1 - 4 353 6762930
BU530423.1 - 4 390
CJ043213.1 - 4 400
Y16430.2 - 3 338

Gene Symbol
Ino80
Entrez Gene ID
68142
Gene Name
INO80 homolog (S. cerevisiae)
Gene Aliases
2310079N15Rik, 4632409L19Rik, Inoc1
Chromosome Location
Chr. 2: 119373042 - 119477629 on Build GRCm38
UniGene ID
Mm.330496
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC061495.1 - 4287

View Full Product Details