Gene Transcripts

Gene Symbol
Ntn4
Entrez Gene ID
57764
Gene Name
netrin 4
Gene Aliases
Chromosome Location
Chr. 10: 93641049 - 93745972 on Build GRCm38
UniGene ID
Mm.483868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021320.3 NP_067295.2 7, 8 1923
GenBank mRNA AF268066.1 - 7, 8 1518
AF281278.1 - 7, 8 1771
BC119182.1 - 7, 8 1605 8733931
BC119184.1 - 7, 8 1605 8733933
CF537120.1 - 3, 4 517

View Full Product Details