Gene Transcripts

Gene Symbol
Robo4
Entrez Gene ID
74144
Gene Name
roundabout homolog 4 (Drosophila)
Gene Aliases
1200012D01Rik, AI593217
Chromosome Location
Chr. 9: 37401902 - 37414023 on Build GRCm38
UniGene ID
Mm.485070
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF536772.1 - 3 461
AK004723.2 - 3 458
AK087355.1 - 3 455
BC071193.1 - 2 151 5060473

View Full Product Details