Gene Transcripts

Gene Symbol
Miip
Entrez Gene ID
28010
Gene Name
migration and invasion inhibitory protein
Gene Aliases
AA553001, AI853621, D4Wsu114e, IIP45
Chromosome Location
Chr. 4: 147860778 - 147868719 on Build GRCm38
UniGene ID
Mm.250504
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025365.2 NP_001020536.2 2, 3 542
GenBank mRNA AK133976.1 - 4, 5 694
AK170224.1 - 2, 3 335
BC017525.1 - 2, 3 285 4910951
BC024069.1 - 2, 3 324 5097088
FJ618908.1 - 2, 3 542

View Full Product Details