Gene Transcripts

Gene Symbol
N4bp2l1
Entrez Gene ID
100637
Gene Name
NEDD4 binding protein 2-like 1
Gene Aliases
2410024N18Rik, AI428195, B230342M21Rik
Chromosome Location
Chr. 5: 150571643 - 150594525 on Build GRCm38
UniGene ID
Mm.279932
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133898.4 NP_598659.3 5 891
GenBank mRNA AK046116.1 - 5 868
AK131647.1 - 5 883
BC019754.1 - 4 853 4007588

View Full Product Details