Gene Transcripts
- Gene Symbol
- Oasl1
- Entrez Gene ID
- 231655
- Gene Name
- 2'-5' oligoadenylate synthetase-like 1
- Gene Aliases
- 7530414C13Rik, oasl9
- Chromosome Location
- Chr. 5: 114923190 - 114937912 on Build GRCm38
- UniGene ID
- Mm.95479
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145209.3 |
NP_660210.1
|
2, 3 |
292 |
|
GenBank mRNA |
AB067533.1 |
- |
1, 2 |
217 |
|
AF426289.1 |
- |
1, 2 |
223 |
|
AK078690.1 |
- |
2, 3 |
293 |
|
AK089111.1 |
- |
1, 2 |
262 |
|
AK149824.1 |
- |
1, 2 |
216 |
|
AK150863.1 |
- |
1, 2 |
346 |
|
AK165578.1 |
- |
1, 2 |
329 |
|
AY057107.1 |
- |
1, 2 |
205 |
|
AY089728.1 |
- |
1, 2 |
217 |
|
BC032152.1 |
- |
1, 2 |
229 |
5068699 |
View Full Product Details