Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_146103.2 | NP_666215.1 | 1 | 1671 | |
GenBank mRNA | AK010846.1 | - | 1 | 1387 | |
AK031130.1 | - | 1 | 1667 | ||
BC020408.1 | - | 1 | 1659 | ||
BC024712.1 | - | 1 | 1658 | 2646072 | |
BC051523.1 | - | 1 | 528 |