Gene Transcripts

Gene Symbol
Olfr1208
Entrez Gene ID
258774
Gene Name
olfactory receptor 1208
Gene Aliases
MOR225-4
Chromosome Location
Chr. 2: 88896669 - 88897595 on Build GRCm38
UniGene ID
Mm.377685
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146778.1 NP_666989.1 1 149
GenBank mRNA BC150697.1 - 1 202

View Full Product Details