Gene Transcripts
- Gene Symbol
- Olfr176
- Entrez Gene ID
- 258995
- Gene Name
- olfactory receptor 176
- Gene Aliases
- GA_x54KRFPKG5P-55043245-55042289, MOR184-8
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.223385
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146993.1 |
NP_667204.1
|
1 |
925 |
|
View Full Product Details