Gene Transcripts
- Gene Symbol
- Vps18
- Entrez Gene ID
- 228545
- Gene Name
- vacuolar protein sorting 18 (yeast)
- Gene Aliases
- 9930024E13Rik
- Chromosome Location
- Chr. 2: 119288742 - 119298453 on Build GRCm38
- UniGene ID
- Mm.294761
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172269.3 |
NP_758473.3
|
2, 3 |
525 |
|
GenBank mRNA |
AK029109.1 |
- |
2, 3 |
502 |
|
AK033333.1 |
- |
2, 3 |
510 |
|
AK036915.1 |
- |
2, 3 |
510 |
|
AK039848.1 |
- |
2, 3 |
510 |
|
AK042471.1 |
- |
2, 3 |
513 |
|
AK044821.1 |
- |
2, 3 |
528 |
|
AK148077.1 |
- |
2, 3 |
514 |
|
AK160689.1 |
- |
2, 3 |
502 |
|
BC036129.1 |
- |
2, 3 |
453 |
4972693 |
BC039176.1 |
- |
2, 3 |
432 |
4221232 |
View Full Product Details