Gene Transcripts

Gene Symbol
NEK6
Entrez Gene ID
10783
Gene Name
NIMA related kinase 6
Gene Aliases
SID6-1512
Chromosome Location
Chr. 9: 124257606 - 124352442 on Build GRCh38
UniGene ID
Hs.197071
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145001.2 NP_001138473.1 3, 4 463
NM_001166167.1 NP_001159639.1 2, 3 245
NM_001166168.1 NP_001159640.1 2, 3 240
NM_001166169.1 NP_001159641.1 2, 3 249
NM_001166170.1 NP_001159642.1 2, 3 247
NM_001166171.1 NP_001159643.1 3, 4 412
NM_014397.5 NP_055212.2 2, 3 298
XM_005251664.1 XP_005251721.1 2, 3 304
XM_006716936.2 XP_006716999.1 2, 3 309
XM_017014217.1 XP_016869706.1 3, 4 472
GenBank mRNA AB026289.1 - 2, 3 229
AF087909.1 - 2, 3 234
AK294614.1 - 2, 3 249
AK313071.1 - 2, 3 199
BC000101.2 - 2, 3 244 3507262
BC004174.2 - 3, 4 416 2958695
BC004209.2 - 3, 4 416 2958695
BC012761.2 - 2, 3 245 3640103
BM083872.1 - 2, 3 245
BP265230.1 - 2, 3 249
BP279761.1 - 2, 3 240
CF994196.1 - 2, 3 325
CR457091.1 - 1, 2 79
CR542222.1 - 1, 2 79

View Full Product Details