Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175088.5 | NP_780297.3 | 3 | 503 | |
GenBank mRNA | AK040675.1 | - | 2 | 215 | |
AK048821.1 | - | 3 | 443 | ||
AK080178.1 | - | 3 | 453 | ||
AK086117.1 | - | 3 | 472 | ||
AK134050.1 | - | 3 | 444 | ||
AK165016.1 | - | 3 | 504 | ||
BB603899.2 | - | 3 | 470 | ||
BC067006.1 | - | 3 | 234 |