Gene Transcripts

Gene Symbol
Heg1
Entrez Gene ID
77446
Gene Name
HEG homolog 1 (zebrafish)
Gene Aliases
4632417D23Rik, 5530401I02Rik, 9530025L16Rik, BC038614, Gm629, OTTMUSG00000027315
Chromosome Location
Chr. 16: 33684430 - 33771576 on Build GRCm38
UniGene ID
Mm.245741
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175256.5 NP_780465.4 15 3267
GenBank mRNA AK043598.1 - 4 1505
BC072651.1 - 7 577
BC076596.1 - 13 2546 30645937

View Full Product Details