Gene Transcripts

Gene Symbol
Adgrb3
Entrez Gene ID
210933
Gene Name
adhesion G protein-coupled receptor B3
Gene Aliases
A830096D10Rik, Bai3
Chromosome Location
Chr. 1: 25067476 - 25830206 on Build GRCm38
UniGene ID
Mm.336569
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175642.4 NP_783573.4 2 566
GenBank mRNA AY168406.1 - 1 305
BC099951.1 - 2 564 30098919

View Full Product Details