gRNA Design Details
- Target DNA Sequence
- GATGAGCGCTCAGGAATCAT
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 32440001 - 32440023 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HLA-DRA
- Entrez Gene ID
-
3122
- Gene Name
- major histocompatibility complex, class II, DR alpha
- Gene Aliases
-
HLA-DRA1, MLRW
- Chromosome Location
-
Chr.6: 32439842 - 32445046 on GRCh38
- UniGene ID
- Hs.520048
- Species
-
Human