gRNA Design Details

Target DNA Sequence
GACACGCACTTATCATTCAG
PAM Sequence
TGG
Target locus
Chr.6: 159991301 - 159991323 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 86.96/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IGF2R
Entrez Gene ID
3482
Gene Name
insulin like growth factor 2 receptor
Gene Aliases
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI
Chromosome Location
Chr.6: 159969099 - 160111504 on GRCh38
UniGene ID
Hs.487062
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000876.3 NP_000867.2 2