gRNA Design Details
- Target DNA Sequence
- CTCTGTGATTTGTGCCTTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 160009074 - 160009096 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.28/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGF2R
- Entrez Gene ID
-
3482
- Gene Name
- insulin like growth factor 2 receptor
- Gene Aliases
-
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI
- Chromosome Location
-
Chr.6: 159969099 - 160111504 on GRCh38
- UniGene ID
- Hs.487062
- Species
-
Human