gRNA Design Details

Target DNA Sequence
ATCTCCCCTCGTGGCCATAT
PAM Sequence
GGG
Target locus
Chr.5: 159326710 - 159326732 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 89.2/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IL12B
Entrez Gene ID
3593
Gene Name
interleukin 12B
Gene Aliases
CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2
Chromosome Location
Chr.5: 159314783 - 159330473 on GRCh38
UniGene ID
Hs.674
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_002187.2 NP_002178.2 2