gRNA Design Details
- Target DNA Sequence
- GTCAAGCAGAATGAGCGCTA
- PAM Sequence
- CGG
- Target locus
-
Chr.8: 17621052 - 17621074 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
93.53/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PDGFRL
- Entrez Gene ID
-
5157
- Gene Name
- platelet derived growth factor receptor like
- Gene Aliases
-
PDGRL, PRLTS
- Chromosome Location
-
Chr.8: 17576433 - 17643133 on GRCh38
- UniGene ID
- Hs.458573
- Species
-
Human