gRNA Design Details
- Target DNA Sequence
- CTAACCCGTTGATCAGCAGA
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 101164783 - 101164805 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- VGF
- Entrez Gene ID
-
7425
- Gene Name
- VGF nerve growth factor inducible
- Gene Aliases
-
SCG7, SgVII
- Chromosome Location
-
Chr.7: 101162509 - 101169956 on GRCh38
- UniGene ID
- Hs.587325
- Species
-
Human